The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177701.2 | NP_001171172.1 | 4 - 5 | 661 | 101 |
NM_006860.4 | NP_006851.1 | 4 - 5 | 658 | 101 | |
XM_006724106.2 | XP_006724169.1 | 5 - 6 | 422 | 101 | |
XM_017028540.1 | XP_016884029.1 | 4 - 5 | 665 | 101 | |
GenBank mRNA | AA809074.1 | - | 3 - 4 | 101 | |
AB209424.1 | - | 4 - 5 | 653 | 101 | |
AL022729.1 | - | 4 - 5 | 602 | 101 | |
BC000566.2 | - | 4 - 5 | 639 | 101 | |
BM711121.1 | - | 4 - 5 | 249 | 101 | |
BT006815.1 | - | 4 - 5 | 235 | 101 | |
CR456558.1 | - | 4 - 5 | 277 | 101 |