The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178562.4 | NP_848657.1 | 5 - 6 | 728 | 112 |
XM_006715960.3 | XP_006716023.1 | 5 - 6 | 556 | 112 | |
GenBank mRNA | AF276891.1 | - | 5 - 6 | 463 | 112 |
AY236849.1 | - | 5 - 6 | 552 | 112 | |
BC044244.1 | - | 5 - 6 | 692 | 112 |