Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX2
Entrez Gene ID
55852
Gene Name
testis expressed 2
Gene Aliases
HT008, TMEM96
Chromosome Location
Chr.17: 64147433 - 64263323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288732.1 NP_001275661.1 3 - 4 2022 86
NM_001288733.1 NP_001275662.1 3 - 4 2014 86
NM_018469.4 NP_060939.3 3 - 4 2022 86
XM_011524998.1 XP_011523300.1 3 - 4 2011 86
XM_011524999.1 XP_011523301.1 3 - 4 1935 86
XM_011525000.1 XP_011523302.1 3 - 4 1992 86
XM_017024846.1 XP_016880335.1 3 - 4 1992 86
GenBank mRNA AB051525.2 - 2 - 3 1841 86
AF163260.1 - 1 - 2 59 86
AK000758.1 - 2 - 3 258 86
AK091156.1 - 2 - 3 1011 86
AK091214.1 - 3 - 4 1939 86
AL832371.1 - 3 - 4 1922 86
BC008896.2 - 2 - 3 1199 86
BC036672.1 - 3 - 4 1993 86
BC040521.1 - 3 - 4 1926 86
BC040522.1 - 3 - 4 1995 86
BC108279.1 - 1 - 2 80 86
CR627433.1 - 3 - 4 2015 86

View Full Product Details