Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX2
Entrez Gene ID
55852
Gene Name
testis expressed 2
Gene Aliases
HT008, TMEM96
Chromosome Location
Chr.17: 64147433 - 64263323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288732.1 NP_001275661.1 9 - 10 3117 157
NM_001288733.1 NP_001275662.1 9 - 10 3109 157
NM_018469.4 NP_060939.3 9 - 10 3138 157
XM_011524998.1 XP_011523300.1 9 - 10 3127 157
XM_011524999.1 XP_011523301.1 9 - 10 3051 157
XM_011525000.1 XP_011523302.1 9 - 10 3102 157
XM_017024846.1 XP_016880335.1 9 - 10 3081 157
GenBank mRNA AB051525.2 - 8 - 9 2936 157
AF163260.1 - 7 - 8 1154 157
AF220182.1 - 5 - 6 672 157
AK000369.1 - 4 - 5 443 157
AK000758.1 - 8 - 9 1353 157
AK091156.1 - 8 - 9 2106 157
AK314316.1 - 4 - 5 388 157
AL832371.1 - 9 - 10 3017 157
AL834251.2 - 9 - 10 3028 157
BC008896.2 - 8 - 9 2293 157
BC033661.1 - 6 - 7 820 157
BC036672.1 - 9 - 10 3088 157
BC040521.1 - 9 - 10 3042 157
BC040522.1 - 9 - 10 3090 157
BC108279.1 - 7 - 8 1175 157
BX648260.1 - 1 - 2 1399 157
CR627433.1 - 9 - 10 3110 157

View Full Product Details