The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015381.6 | NP_056196.2 | 1 - 2 | 269 | 67 |
GenBank mRNA | AK123443.1 | - | 2 - 3 | 208 | 67 |
AK125256.1 | - | 1 - 2 | 269 | 67 | |
AY325118.1 | - | 1 - 2 | 243 | 67 |