Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRAME
Entrez Gene ID
23532
Gene Name
preferentially expressed antigen in melanoma
Gene Aliases
CT130, MAPE, OIP-4, OIP4
Chromosome Location
Chr.22: 22547696 - 22559765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006115.4 NP_006106.1 1 - 2 188 125
NM_206954.2 NP_996837.1 1 - 2 188 104
GenBank mRNA BQ432004.1 - 1 - 2 111 104
U65011.1 - 1 - 2 102 125

View Full Product Details