Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NPHP3
- Entrez Gene ID
-
27031
- Gene Name
- nephronophthisis 3 (adolescent)
- Gene Aliases
-
CFAP31, MKS7, NPH3, RHPD, RHPD1, SLSN3
- Chromosome Location
-
Chr.3: 132680609 - 132722459 on Build GRCh38
- Gene Symbol
- NPHP3-ACAD11
- Entrez Gene ID
-
100532724
- Gene Name
- NPHP3-ACAD11 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 132558138 - 132722459 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037804.1 |
|
21 - 22 |
3213 |
144 |
GenBank mRNA |
AB082531.1 |
- |
21 - 22 |
3213 |
144 |
View Full Product Details