The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153240.4 | NP_694972.3 | 12 - 13 | 1984 | 84 |
GenBank mRNA | AK055253.1 | - | 7 - 8 | 927 | 84 |
AK092910.1 | - | 2 - 3 | 263 | 84 | |
AL832863.1 | - | 2 - 3 | 218 | 84 | |
AY257864.1 | - | 12 - 13 | 1913 | 84 |