The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252294.1 | NP_001239223.1 | 4 - 5 | 1216 | 61 |
NM_012391.2 | NP_036523.1 | 5 - 6 | 1264 | 61 | |
XM_005248988.4 | XP_005249045.2 | 5 - 6 | 1956 | 61 | |
GenBank mRNA | AB031549.1 | - | 5 - 6 | 1235 | 61 |
AF071538.1 | - | 5 - 6 | 1245 | 61 | |
AK301543.1 | - | 4 - 5 | 1216 | 61 | |
BC021299.2 | - | 5 - 6 | 1241 | 61 |