The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004249.3 | NP_004240.2 | 6 - 7 | 787 | 109 |
GenBank mRNA | AF498955.1 | - | 6 - 7 | 572 | 109 |
X94703.1 | - | 6 - 7 | 572 | 109 |