The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000730.2 | NP_000721.1 | 3 - 4 | 783 | 94 |
GenBank mRNA | AK313978.1 | - | 3 - 4 | 825 | 94 |
AY322549.1 | - | 3 - 4 | 630 | 94 | |
BC074987.2 | - | 3 - 4 | 678 | 94 | |
L13605.1 | - | 3 - 4 | 701 | 94 | |
L19315.1 | - | 3 - 4 | 783 | 94 |