The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001430.4 | NP_001421.2 | 6 - 7 | 1284 | 66 |
XM_011532698.2 | XP_011531000.1 | 6 - 7 | 1042 | 66 | |
GenBank mRNA | BC051338.1 | - | 6 - 7 | 1089 | 66 |
U51626.1 | - | 6 - 7 | 806 | 66 | |
U81984.1 | - | 6 - 7 | 923 | 66 |