Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIT
Entrez Gene ID
5212
Gene Name
vitrin
Gene Aliases
-
Chromosome Location
Chr.2: 36696685 - 36814794 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177969.1 NP_001171440.1 8 - 9 1030 71
NM_001177970.1 NP_001171441.1 8 - 9 1030 71
NM_053276.3 NP_444506.2 8 - 9 1030 71
XM_011532905.1 XP_011531207.1 7 - 8 735 71
XM_017004327.1 XP_016859816.1 6 - 7 933 71
GenBank mRNA AK056772.1 - 8 - 9 949 71
AY358338.1 - 8 - 9 915 71
BC128260.1 - 8 - 9 843 71

View Full Product Details