Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STEAP4
Entrez Gene ID
79689
Gene Name
STEAP4 metalloreductase
Gene Aliases
STAMP2, TIARP, TNFAIP9
Chromosome Location
Chr.7: 88276429 - 88306913 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205315.1 NP_001192244.1 3 - 4 684 61
NM_024636.3 NP_078912.2 2 - 3 585 61
GenBank mRNA AF423422.1 - 3 - 4 567 61
AF423423.1 - 3 - 4 567 61
AK026806.1 - 2 - 3 564 61
AK307644.1 - 2 - 3 563 61

View Full Product Details