The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205315.1 | NP_001192244.1 | 5 - 6 | 1371 | 75 |
NM_001205316.1 | NP_001192245.1 | 3 - 4 | 744 | 75 | |
NM_024636.3 | NP_078912.2 | 4 - 5 | 1272 | 75 | |
GenBank mRNA | AF423422.1 | - | 5 - 6 | 1254 | 75 |
AK026806.1 | - | 4 - 5 | 1253 | 75 | |
AL832955.1 | - | 2 - 3 | 363 | 75 | |
AL833044.1 | - | 2 - 3 | 177 | 75 | |
AL833671.1 | - | 1 - 2 | 93 | 75 | |
BC020600.1 | - | 3 - 4 | 719 | 75 |