The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040428.3 | NP_001035518.1 | 8 - 9 | 1159 | 164 |
NM_018418.4 | NP_060888.2 | 9 - 10 | 1255 | 164 | |
XM_005267851.1 | XP_005267908.1 | 9 - 10 | 1229 | 164 | |
XM_005267852.1 | XP_005267909.1 | 8 - 9 | 1158 | 164 | |
XM_005267854.1 | XP_005267911.1 | 10 - 11 | 1291 | 164 | |
XM_005267855.1 | XP_005267912.1 | 9 - 10 | 1224 | 164 | |
XM_006720204.1 | XP_006720267.1 | 9 - 10 | 1229 | 164 | |
XM_006720205.1 | XP_006720268.1 | 9 - 10 | 1229 | 164 | |
XM_011536951.1 | XP_011535253.1 | 9 - 10 | 1423 | 164 | |
XM_011536952.1 | XP_011535254.1 | 7 - 8 | 1041 | 164 | |
XM_011536953.1 | XP_011535255.1 | 8 - 9 | 1362 | 164 | |
XM_017021452.1 | XP_016876941.1 | 9 - 10 | 1420 | 164 | |
XM_017021453.1 | XP_016876942.1 | 9 - 10 | 1389 | 164 | |
XM_017021454.1 | XP_016876943.1 | 10 - 11 | 1288 | 164 | |
XM_017021455.1 | XP_016876944.1 | 9 - 10 | 1221 | 164 | |
XM_017021456.1 | XP_016876945.1 | 10 - 11 | 1234 | 164 | |
XM_017021457.1 | XP_016876946.1 | 8 - 9 | 1359 | 164 | |
GenBank mRNA | AF144487.3 | - | 9 - 10 | 1235 | 164 |
AF144488.2 | - | 7 - 8 | 963 | 164 | |
AK055864.1 | - | 8 - 9 | 2827 | 164 | |
AK225746.1 | - | 10 - 11 | 1229 | 164 | |
AK290631.1 | - | 8 - 9 | 1130 | 164 | |
AL136604.1 | - | 8 - 9 | 1139 | 164 | |
BC008656.1 | - | 4 - 5 | 687 | 164 | |
BC090875.1 | - | 9 - 10 | 1542 | 164 | |
CR533563.1 | - | 8 - 9 | 984 | 164 | |
CR936777.1 | - | 9 - 10 | 1201 | 164 | |
EU668353.1 | - | 9 - 10 | 1231 | 164 |