Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBRS
Entrez Gene ID
64319
Gene Name
fibrosin
Gene Aliases
FBS, FBS1
Chromosome Location
Chr.16: 30658142 - 30670810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105079.2 NP_001098549.2 10 - 11 1710 140
XM_011545917.2 XP_011544219.1 9 - 10 3057 140
XM_011545919.2 XP_011544221.1 8 - 9 3020 140
GenBank mRNA AK022551.1 - 1 - 2 115 140
AK298229.1 - 3 - 4 273 140
BG775600.1 - 4 - 5 459 140
BQ276414.1 - 3 - 4 278 140
DA527487.1 - 2 - 3 134 140
DC408959.1 - 3 - 4 361 140
DR761499.1 - 3 - 4 207 140

View Full Product Details