Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDH6
Entrez Gene ID
1004
Gene Name
cadherin 6
Gene Aliases
CAD6, KCAD
Chromosome Location
Chr.5: 31193655 - 31329146 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004932.3 NP_004923.1 10 - 11 1990 107
XM_011513921.2 XP_011512223.1 10 - 11 1809 107
XM_017008910.1 XP_016864399.1 9 - 10 1760 107
XM_017008911.1 XP_016864400.1 10 - 11 1947 107
GenBank mRNA AK291290.1 - 10 - 11 1956 107
BC000019.2 - 10 - 11 1895 107
D31784.1 - 9 - 10 1750 107

View Full Product Details