The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004863.3 | NP_004854.1 | 8 - 9 | 1358 | 71 |
XM_011537384.2 | XP_011535686.1 | 8 - 9 | 1254 | 71 | |
GenBank mRNA | AB011098.1 | - | 8 - 9 | 1358 | 71 |
BC005123.2 | - | 8 - 9 | 1259 | 71 | |
U15555.1 | - | 7 - 8 | 994 | 71 | |
Y08686.1 | - | 8 - 9 | 1218 | 71 |