The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290331.1 | NP_001277260.1 | 9 - 10 | 784 | 86 |
NM_024898.3 | NP_079174.2 | 11 - 12 | 893 | 86 | |
XM_006722905.2 | XP_006722968.2 | 10 - 11 | 763 | 86 | |
XM_006722906.3 | XP_006722969.1 | 11 - 12 | 822 | 86 | |
XM_011528318.2 | XP_011526620.1 | 10 - 11 | 813 | 86 | |
GenBank mRNA | AK026410.1 | - | 11 - 12 | 872 | 86 |
AK055820.1 | - | 1 - 2 | 380 | 86 | |
AK225623.1 | - | 11 - 12 | 872 | 86 | |
AK303226.1 | - | 9 - 10 | 763 | 86 | |
BC033437.1 | - | 11 - 12 | 860 | 86 |