The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290331.1 | NP_001277260.1 | 15 - 16 | 1292 | 83 |
NM_024898.3 | NP_079174.2 | 17 - 18 | 1401 | 83 | |
XM_006722905.2 | XP_006722968.2 | 15 - 16 | 1145 | 83 | |
XM_006722906.3 | XP_006722969.1 | 17 - 18 | 1330 | 83 | |
XM_011528318.2 | XP_011526620.1 | 16 - 17 | 1321 | 83 | |
GenBank mRNA | AK026410.1 | - | 17 - 18 | 1380 | 83 |
AK055820.1 | - | 6 - 7 | 1008 | 83 | |
AK303226.1 | - | 15 - 16 | 1271 | 83 | |
AL713770.1 | - | 15 - 16 | 1521 | 83 | |
BC033437.1 | - | 17 - 18 | 1368 | 83 | |
BX647304.1 | - | 5 - 6 | 1031 | 83 |