The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024321.4 | NP_077297.2 | 4 - 5 | 576 | 77 |
XM_005259257.2 | XP_005259314.1 | 4 - 5 | 571 | 77 | |
GenBank mRNA | BC004204.2 | - | 4 - 5 | 514 | 77 |