Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
Chromosome Location
Chr.11: 8612037 - 8683217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014818.1 NP_055633.1 16 - 17 3341 70
XM_006718397.2 XP_006718460.1 21 - 22 4279 70
XM_006718398.3 XP_006718461.1 17 - 18 4891 70
XM_011520504.1 XP_011518806.1 22 - 23 4322 70
XM_011520507.2 XP_011518809.1 21 - 22 4243 70
XM_011520508.1 XP_011518810.1 21 - 22 4279 70
XM_011520510.1 XP_011518812.1 21 - 22 4195 70
XM_011520511.1 XP_011518813.1 21 - 22 4168 70
XM_011520512.1 XP_011518814.1 17 - 18 3543 70
XM_011520513.1 XP_011518815.1 17 - 18 3541 70
XM_011520514.2 XP_011518816.1 17 - 18 3497 70
XM_011520515.1 XP_011518817.1 18 - 19 3710 70
XM_011520516.1 XP_011518818.1 18 - 19 3710 70
XM_011520517.2 XP_011518819.1 18 - 19 5032 70
XM_011520518.1 XP_011518820.1 18 - 19 3680 70
XM_011520519.1 XP_011518821.1 17 - 18 3568 70
XM_011520522.2 XP_011518824.1 16 - 17 4746 70
XM_011520523.2 XP_011518825.1 17 - 18 5628 70
XM_011520526.2 XP_011518828.1 9 - 10 2597 70
XM_011520527.1 XP_011518829.1 9 - 10 1865 70
XM_017018629.1 XP_016874118.1 21 - 22 4672 70
XM_017018630.1 XP_016874119.1 16 - 17 3634 70
XM_017018631.1 XP_016874120.1 16 - 17 3786 70
GenBank mRNA AB002296.2 - 15 - 16 3595 70

View Full Product Details