Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
Chromosome Location
Chr.11: 8612037 - 8683217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014818.1 NP_055633.1 19 - 20 3785 73
XM_006718397.2 XP_006718460.1 24 - 25 4723 73
XM_006718398.3 XP_006718461.1 20 - 21 5335 73
XM_011520504.1 XP_011518806.1 25 - 26 4766 73
XM_011520507.2 XP_011518809.1 24 - 25 4687 73
XM_011520509.1 XP_011518811.1 23 - 24 4648 73
XM_011520510.1 XP_011518812.1 24 - 25 4639 73
XM_011520511.1 XP_011518813.1 24 - 25 4612 73
XM_011520512.1 XP_011518814.1 20 - 21 3987 73
XM_011520513.1 XP_011518815.1 20 - 21 3985 73
XM_011520514.2 XP_011518816.1 20 - 21 3941 73
XM_011520515.1 XP_011518817.1 21 - 22 4154 73
XM_011520516.1 XP_011518818.1 21 - 22 4154 73
XM_011520517.2 XP_011518819.1 21 - 22 5476 73
XM_011520518.1 XP_011518820.1 21 - 22 4124 73
XM_011520519.1 XP_011518821.1 20 - 21 4012 73
XM_011520522.2 XP_011518824.1 19 - 20 5190 73
XM_011520523.2 XP_011518825.1 20 - 21 6072 73
XM_011520526.2 XP_011518828.1 12 - 13 3041 73
XM_011520527.1 XP_011518829.1 12 - 13 2309 73
XM_017018629.1 XP_016874118.1 24 - 25 5116 73
XM_017018630.1 XP_016874119.1 19 - 20 4078 73
XM_017018631.1 XP_016874120.1 19 - 20 4230 73
GenBank mRNA AB002296.2 - 18 - 19 4039 73

View Full Product Details