Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
Chromosome Location
Chr.11: 8612037 - 8683217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014818.1 NP_055633.1 2 - 3 201 66
XM_006718397.2 XP_006718460.1 7 - 8 1046 66
XM_006718398.3 XP_006718461.1 3 - 4 1658 66
XM_011520504.1 XP_011518806.1 8 - 9 1089 66
XM_011520507.2 XP_011518809.1 7 - 8 1010 66
XM_011520508.1 XP_011518810.1 7 - 8 1046 66
XM_011520509.1 XP_011518811.1 7 - 8 1046 66
XM_011520510.1 XP_011518812.1 7 - 8 1046 66
XM_011520511.1 XP_011518813.1 7 - 8 935 66
XM_011520512.1 XP_011518814.1 3 - 4 310 66
XM_011520513.1 XP_011518815.1 3 - 4 308 66
XM_011520514.2 XP_011518816.1 3 - 4 264 66
XM_011520515.1 XP_011518817.1 4 - 5 477 66
XM_011520516.1 XP_011518818.1 4 - 5 477 66
XM_011520517.2 XP_011518819.1 4 - 5 1799 66
XM_011520518.1 XP_011518820.1 4 - 5 447 66
XM_011520519.1 XP_011518821.1 3 - 4 335 66
XM_011520522.2 XP_011518824.1 2 - 3 1513 66
XM_011520523.2 XP_011518825.1 3 - 4 2395 66
XM_011520524.1 XP_011518826.1 7 - 8 1046 66
XM_011520525.1 XP_011518827.1 7 - 8 1046 66
XM_017018629.1 XP_016874118.1 7 - 8 1439 66
XM_017018630.1 XP_016874119.1 2 - 3 401 66
XM_017018631.1 XP_016874120.1 2 - 3 553 66
GenBank mRNA AB002296.2 - 2 - 3 201 66
AK308310.1 - 4 - 5 447 66

View Full Product Details