The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099403.1 | NP_001092873.1 | 2 - 3 | 487 | 125 |
NM_020226.3 | NP_064611.3 | 8 - 9 | 1455 | 125 | |
XM_005263144.2 | XP_005263201.1 | 7 - 8 | 1271 | 125 | |
XM_005263145.2 | XP_005263202.1 | 5 - 6 | 1044 | 125 | |
XM_005263146.4 | XP_005263203.1 | 6 - 7 | 1075 | 125 | |
XM_011532133.2 | XP_011530435.1 | 6 - 7 | 1492 | 125 | |
XM_011532135.2 | XP_011530437.1 | 5 - 6 | 1351 | 125 | |
XM_011532140.2 | XP_011530442.1 | 6 - 7 | 1060 | 125 | |
XM_011532141.2 | XP_011530443.1 | 4 - 5 | 893 | 125 | |
XM_017008468.1 | XP_016863957.1 | 6 - 7 | 943 | 125 | |
XM_017008469.1 | XP_016863958.1 | 5 - 6 | 1096 | 125 | |
XM_017008470.1 | XP_016863959.1 | 6 - 7 | 943 | 125 | |
GenBank mRNA | AF275815.1 | - | 8 - 9 | 1457 | 125 |
AK292137.1 | - | 2 - 3 | 483 | 125 | |
BC027929.1 | - | 2 - 3 | 473 | 125 | |
BC058908.1 | - | 6 - 7 | 1063 | 125 | |
BC071584.1 | - | 2 - 3 | 487 | 125 | |
BP219935.1 | - | 2 - 3 | 487 | 125 |