The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014865.3 | NP_055680.3 | 22 - 23 | 2956 | 73 |
GenBank mRNA | AK125155.1 | - | 19 - 20 | 2611 | 73 |
AK128354.1 | - | 22 - 23 | 2868 | 73 | |
AK303310.1 | - | 21 - 22 | 2931 | 73 | |
BC028182.1 | - | 22 - 23 | 2968 | 73 | |
D63880.1 | - | 22 - 23 | 3709 | 73 |