The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014865.3 | NP_055680.3 | 27 - 28 | 3614 | 69 |
GenBank mRNA | AK022511.1 | - | 27 - 28 | 3614 | 69 |
AK128354.1 | - | 27 - 28 | 3526 | 69 | |
AK303310.1 | - | 26 - 27 | 3589 | 69 | |
BC028182.1 | - | 27 - 28 | 3626 | 69 | |
D63880.1 | - | 27 - 28 | 4367 | 69 |