Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG6
Entrez Gene ID
55200
Gene Name
pleckstrin homology and RhoGEF domain containing G6
Gene Aliases
MyoGEF
Chromosome Location
Chr.12: 6310436 - 6328506 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144856.1 NP_001138328.1 13 - 14 1761 100
NM_001144857.1 NP_001138329.1 12 - 13 1929 100
NM_018173.3 NP_060643.2 13 - 14 1679 100
XM_005253704.4 XP_005253761.1 13 - 14 1776 100
XM_006718985.3 XP_006719048.1 13 - 14 1734 100
XM_011520967.2 XP_011519269.1 13 - 14 1651 100
XM_017019555.1 XP_016875044.1 13 - 14 1776 100
GenBank mRNA AK001527.1 - 13 - 14 1680 100
AK023741.1 - 13 - 14 1761 100
AK095373.1 - 12 - 13 1929 100
BC104751.1 - 13 - 14 1615 100
BC105961.1 - 13 - 14 1615 100
BC109095.2 - 13 - 14 1615 100

View Full Product Details