The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144856.1 | NP_001138328.1 | 15 - 16 | 2589 | 92 |
NM_001144857.1 | NP_001138329.1 | 14 - 15 | 2757 | 92 | |
NM_018173.3 | NP_060643.2 | 15 - 16 | 2507 | 92 | |
XM_005253704.4 | XP_005253761.1 | 15 - 16 | 2604 | 92 | |
XM_006718985.3 | XP_006719048.1 | 15 - 16 | 2562 | 92 | |
XM_011520967.2 | XP_011519269.1 | 15 - 16 | 2479 | 92 | |
GenBank mRNA | AF289613.1 | - | 3 - 4 | 1049 | 92 |
AK001527.1 | - | 15 - 16 | 2508 | 92 | |
AK023741.1 | - | 15 - 16 | 2589 | 92 | |
AK095373.1 | - | 14 - 15 | 2757 | 92 | |
AW001611.1 | - | 1 - 2 | 92 | ||
BC104751.1 | - | 15 - 16 | 2443 | 92 | |
BC105961.1 | - | 15 - 16 | 2443 | 92 | |
BC109095.2 | - | 15 - 16 | 2443 | 92 | |
BU754580.1 | - | 1 - 2 | 92 |