Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHG6
Entrez Gene ID
55200
Gene Name
pleckstrin homology and RhoGEF domain containing G6
Gene Aliases
MyoGEF
Chromosome Location
Chr.12: 6310436 - 6328506 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144856.1 NP_001138328.1 6 - 7 837 96
NM_001144857.1 NP_001138329.1 5 - 6 1005 96
NM_018173.3 NP_060643.2 6 - 7 755 96
XM_005253704.4 XP_005253761.1 6 - 7 852 96
XM_006718985.3 XP_006719048.1 6 - 7 852 96
XM_011520967.2 XP_011519269.1 6 - 7 727 96
XM_017019555.1 XP_016875044.1 6 - 7 852 96
GenBank mRNA AK001527.1 - 6 - 7 756 96
AK023741.1 - 6 - 7 837 96
AK095373.1 - 5 - 6 1005 96
BC104751.1 - 6 - 7 691 96
BC105961.1 - 6 - 7 691 96
BC109095.2 - 6 - 7 691 96

View Full Product Details