The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105669.2 | NP_001099139.2 | 4 - 5 | 1074 | 132 |
XM_011509253.2 | XP_011507555.1 | 3 - 4 | 456 | 132 | |
XM_011509254.2 | XP_011507556.1 | 3 - 4 | 456 | 132 | |
XM_011509255.2 | XP_011507557.1 | 3 - 4 | 456 | 132 | |
GenBank mRNA | BQ216668.1 | - | 2 - 3 | 338 | 132 |