The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031271.3 | NP_112561.2 | 1 - 2 | 7012 | 104 |
XM_005273575.3 | XP_005273632.1 | 8 - 9 | 8380 | 104 | |
XM_005273576.3 | XP_005273633.1 | 5 - 6 | 7942 | 104 | |
XM_006716369.3 | XP_006716432.1 | 7 - 8 | 8272 | 104 | |
XM_011544588.2 | XP_011542890.1 | 7 - 8 | 8236 | 104 | |
XM_011544589.2 | XP_011542891.1 | 8 - 9 | 8345 | 104 | |
XM_011544590.2 | XP_011542892.1 | 8 - 9 | 8611 | 104 | |
XM_011544591.2 | XP_011542893.1 | 7 - 8 | 8273 | 104 | |
XM_011544592.2 | XP_011542894.1 | 7 - 8 | 8211 | 104 | |
XM_011544593.2 | XP_011542895.1 | 5 - 6 | 9168 | 104 | |
GenBank mRNA | AF285605.1 | - | 1 - 2 | 7012 | 104 |
BC140934.1 | - | 1 - 2 | 7087 | 104 |