Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MACROD2
Entrez Gene ID
140733
Gene Name
MACRO domain containing 2
Gene Aliases
C20orf133
Chromosome Location
Chr.20: 13995476 - 16053197 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033087.1 NP_001028259.1 12 - 13 981 77
XM_017027675.1 XP_016883164.1 9 - 10 625 77
XM_017027677.1 XP_016883166.1 16 - 17 1512 77
GenBank mRNA BC018687.1 - 12 - 13 984 77
BC035876.1 - 4 - 5 425 77
EF159161.1 - - 1224 77

View Full Product Details