Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MACROD2
Entrez Gene ID
140733
Gene Name
MACRO domain containing 2
Gene Aliases
C20orf133
Chromosome Location
Chr.20: 13995476 - 16053197 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC035876.1 - 5 - 6 502 69

View Full Product Details