The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033087.1 | NP_001028259.1 | 7 - 8 | 592 | 67 |
NM_080676.5 | NP_542407.2 | 11 - 12 | 1099 | 67 | |
XM_017027675.1 | XP_016883164.1 | 4 - 5 | 236 | 67 | |
XM_017027676.1 | XP_016883165.1 | 4 - 5 | 236 | 67 | |
XM_017027677.1 | XP_016883166.1 | 11 - 12 | 1123 | 67 | |
GenBank mRNA | AK125899.1 | - | 4 - 5 | 326 | 67 |
AK131348.1 | - | 11 - 12 | 1229 | 67 | |
BC018687.1 | - | 7 - 8 | 595 | 67 | |
EF159161.1 | - | - | 835 | 67 |