The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033087.1 | NP_001028259.1 | 8 - 9 | 670 | 88 |
NM_080676.5 | NP_542407.2 | 12 - 13 | 1177 | 88 | |
XM_017027675.1 | XP_016883164.1 | 5 - 6 | 314 | 88 | |
XM_017027676.1 | XP_016883165.1 | 5 - 6 | 314 | 88 | |
XM_017027677.1 | XP_016883166.1 | 12 - 13 | 1201 | 88 | |
GenBank mRNA | AK125899.1 | - | 5 - 6 | 404 | 88 |
AK131348.1 | - | 12 - 13 | 1307 | 88 | |
BC018687.1 | - | 8 - 9 | 673 | 88 | |
EF159161.1 | - | - | 913 | 88 |