The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130107.1 | NP_001123579.1 | 2 - 3 | 570 | 62 |
NM_005552.4 | NP_005543.2 | 2 - 3 | 570 | 62 | |
NM_182923.3 | NP_891553.2 | 2 - 3 | 570 | 62 | |
GenBank mRNA | AB451299.1 | - | 1 - 2 | 250 | 62 |
AB451431.1 | - | 1 - 2 | 250 | 62 | |
AF527736.1 | - | 2 - 3 | 520 | 62 | |
AK294508.1 | - | 2 - 3 | 555 | 62 | |
AY180164.2 | - | 2 - 3 | 517 | 62 | |
BC008881.2 | - | 2 - 3 | 531 | 62 | |
L04733.1 | - | 2 - 3 | 526 | 62 |