The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142434.1 | NP_001135906.1 | 8 - 9 | 2099 | 96 |
NM_012215.3 | NP_036347.1 | 9 - 10 | 2258 | 96 | |
XM_017015585.1 | XP_016871074.1 | 10 - 11 | 2635 | 96 | |
XM_017015586.1 | XP_016871075.1 | 6 - 7 | 1422 | 96 | |
GenBank mRNA | AB014579.2 | - | 9 - 10 | 1820 | 96 |
AF036144.2 | - | 9 - 10 | 2210 | 96 | |
AF307332.1 | - | 9 - 10 | 2210 | 96 | |
AK002091.1 | - | 1 - 2 | 610 | 96 | |
BC001343.1 | - | 1 - 2 | 281 | 96 | |
BC039583.2 | - | 8 - 9 | 96 |