The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000478.5 | NP_000469.3 | 2 - 3 | 326 | 71 |
XM_005245818.1 | XP_005245875.1 | 2 - 3 | 296 | 71 | |
XM_006710546.2 | XP_006710609.1 | 2 - 3 | 210 | 71 | |
GenBank mRNA | AB011406.1 | - | 2 - 3 | 238 | 71 |
AB209814.1 | - | 2 - 3 | 298 | 71 | |
AK293184.1 | - | 2 - 3 | 244 | 71 | |
AK312667.1 | - | 2 - 3 | 258 | 71 | |
BC021289.2 | - | 2 - 3 | 250 | 71 | |
BC066116.1 | - | 2 - 3 | 246 | 71 | |
BC090861.1 | - | 2 - 3 | 239 | 71 | |
BC110909.1 | - | 2 - 3 | 226 | 71 | |
BC126165.1 | - | 1 - 2 | 175 | 71 | |
BC136325.1 | - | 1 - 2 | 175 | 71 | |
CN425768.1 | - | 2 - 3 | 317 | 71 | |
DA574668.1 | - | 2 - 3 | 313 | 71 | |
DA625627.1 | - | 2 - 3 | 281 | 71 | |
DA631560.1 | - | 2 - 3 | 285 | 71 | |
HY052024.1 | - | 2 - 3 | 327 | 71 |