The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017770.3 | NP_060240.3 | 2 - 3 | 229 | 104 |
GenBank mRNA | BC050278.1 | - | 2 - 3 | 229 | 104 |
BC060809.1 | - | 2 - 3 | 180 | 104 |