Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGI1
Entrez Gene ID
9223
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location
Chr.3: 65353525 - 66038836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033057.1 NP_001028229.1 7 - 8 1606 101
NM_004742.2 NP_004733.2 7 - 8 1606 101
NM_015520.1 NP_056335.1 7 - 8 1606 101
XM_005265563.1 XP_005265620.1 7 - 8 1614 101
XM_005265564.1 XP_005265621.1 7 - 8 1608 101
XM_005265565.1 XP_005265622.1 7 - 8 1614 101
XM_005265566.1 XP_005265623.1 7 - 8 1608 101
XM_005265570.1 XP_005265627.1 7 - 8 1614 101
XM_005265571.1 XP_005265628.1 7 - 8 1608 101
XM_005265574.3 XP_005265631.1 7 - 8 1608 101
XM_006713407.1 XP_006713470.1 7 - 8 1614 101
XM_006713408.1 XP_006713471.1 7 - 8 1611 101
XM_006713409.1 XP_006713472.1 7 - 8 1611 101
XM_006713411.1 XP_006713474.1 7 - 8 1614 101
XM_006713412.1 XP_006713475.1 7 - 8 1608 101
XM_006713414.3 XP_006713477.1 7 - 8 1614 101
XM_017007497.1 XP_016862986.1 7 - 8 1611 101
XM_017007498.1 XP_016862987.1 7 - 8 1608 101
XM_017007499.1 XP_016862988.1 7 - 8 1969 101
XM_017007500.1 XP_016862989.1 7 - 8 1962 101
XM_017007501.1 XP_016862990.1 7 - 8 1611 101
XM_017007502.1 XP_016862991.1 7 - 8 1611 101
XM_017007503.1 XP_016862992.1 7 - 8 1614 101
XM_017007504.1 XP_016862993.1 7 - 8 1611 101
XM_017007505.1 XP_016862994.1 7 - 8 1611 101
XM_017007506.1 XP_016862995.1 7 - 8 1608 101
XM_017007507.1 XP_016862996.1 6 - 7 772 101
XM_017007508.1 XP_016862997.1 6 - 7 772 101
GenBank mRNA AB010894.1 - 7 - 8 1322 101
AF401654.1 - 7 - 8 1080 101
AF401655.1 - 7 - 8 1080 101
AF401656.1 - 7 - 8 1080 101
AK289803.1 - 7 - 8 1175 101
AK310365.1 - 12 - 13 1607 101
KC170987.1 - 7 - 8 1080 101
U80754.1 - 5 - 6 633 101

View Full Product Details