Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MAGI1
- Entrez Gene ID
-
9223
- Gene Name
- membrane associated guanylate kinase, WW and PDZ domain containing 1
- Gene Aliases
-
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
- Chromosome Location
-
Chr.3: 65353525 - 66038836 on Build GRCh38
View Full Product Details