The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146688.1 | NP_001140160.1 | 21 - 22 | 3376 | 129 |
NM_018433.5 | NP_060903.2 | 21 - 22 | 3645 | 129 | |
XM_006712051.3 | XP_006712114.1 | 20 - 21 | 3343 | 129 | |
XM_017004493.1 | XP_016859982.1 | 21 - 22 | 3664 | 129 | |
XM_017004494.1 | XP_016859983.1 | 22 - 23 | 3487 | 129 | |
GenBank mRNA | AB018285.2 | - | 21 - 22 | 3408 | 129 |
AK074314.1 | - | 7 - 8 | 1042 | 129 | |
AK304276.1 | - | 19 - 20 | 3162 | 129 | |
AL832150.1 | - | 18 - 19 | 5324 | 129 | |
BC038297.2 | - | 21 - 22 | 3422 | 129 | |
BX640698.1 | - | 21 - 22 | 3446 | 129 | |
BX647271.1 | - | 21 - 22 | 3414 | 129 | |
BX647752.1 | - | 21 - 22 | 3474 | 129 | |
BX648841.1 | - | 21 - 22 | 3377 | 129 | |
CR749581.1 | - | 21 - 22 | 3413 | 129 |