The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146688.1 | NP_001140160.1 | 9 - 10 | 1066 | 116 |
NM_018433.5 | NP_060903.2 | 9 - 10 | 1335 | 116 | |
XM_006712051.3 | XP_006712114.1 | 8 - 9 | 1038 | 116 | |
XM_017004493.1 | XP_016859982.1 | 9 - 10 | 1354 | 116 | |
XM_017004494.1 | XP_016859983.1 | 10 - 11 | 1177 | 116 | |
GenBank mRNA | AB018285.2 | - | 9 - 10 | 1098 | 116 |
AK304276.1 | - | 7 - 8 | 852 | 116 | |
AL832150.1 | - | 9 - 10 | 1101 | 116 | |
BC038297.2 | - | 9 - 10 | 1112 | 116 | |
BX640698.1 | - | 9 - 10 | 1136 | 116 | |
BX647271.1 | - | 9 - 10 | 1104 | 116 | |
BX647752.1 | - | 9 - 10 | 1164 | 116 | |
BX648841.1 | - | 9 - 10 | 1067 | 116 | |
BX649109.1 | - | 9 - 10 | 1105 | 116 | |
CR749581.1 | - | 9 - 10 | 1103 | 116 |