The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130010.2 | NP_001123482.1 | 2 - 3 | 405 | 74 |
NM_001290232.1 | NP_001277161.1 | 2 - 3 | 96 | 74 | |
NM_001290233.1 | NP_001277162.1 | 2 - 3 | 405 | 74 | |
NM_001321757.1 | NP_001308686.1 | 2 - 3 | 405 | 74 | |
NM_001321759.1 | NP_001308688.1 | 2 - 3 | 405 | 74 | |
NM_001321760.1 | NP_001308689.1 | 2 - 3 | 405 | 74 | |
NM_001321761.1 | NP_001308690.1 | 2 - 3 | 405 | 74 | |
NM_032499.5 | NP_115888.1 | 3 - 4 | 151 | 74 | |
XM_011522111.1 | XP_011520413.1 | 2 - 3 | 397 | 74 | |
XM_017022676.1 | XP_016878165.1 | 2 - 3 | 397 | 74 | |
XM_017022677.1 | XP_016878166.1 | 2 - 3 | 397 | 74 | |
GenBank mRNA | AF114263.1 | - | 3 - 4 | 126 | 74 |
AK026504.1 | - | 2 - 3 | 374 | 74 | |
AK315446.1 | - | 2 - 3 | 65 | 74 | |
BC006254.2 | - | 2 - 3 | 164 | 74 | |
DA467314.1 | - | 2 - 3 | 397 | 74 | |
DA534917.1 | - | 1 - 2 | 59 | 74 |