Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLF14
Entrez Gene ID
136259
Gene Name
Kruppel like factor 14
Gene Aliases
BTEB5
Chromosome Location
Chr.7: 130732554 - 130734033 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138693.3 NP_619638.2 1 - 1 68
GenBank mRNA AA455939.1 - 1 - 1 168 68
AF490374.1 - 1 - 1 68

View Full Product Details