Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
-
Chromosome Location
Chr.10: 101845599 - 102056237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024541.2 NP_078817.2 20 - 21 1597 98
XM_011540151.2 XP_011538453.1 20 - 21 1642 98
XM_011540152.2 XP_011538454.1 19 - 20 1529 98
XM_011540153.2 XP_011538455.1 19 - 20 1493 98
XM_011540154.2 XP_011538456.1 18 - 19 1427 98
XM_011540156.2 XP_011538458.1 13 - 14 1561 98
XM_011540157.2 XP_011538459.1 19 - 20 1546 98
XM_017016635.1 XP_016872124.1 20 - 21 1595 98
XM_017016636.1 XP_016872125.1 20 - 21 1595 98
GenBank mRNA AK023176.1 - 3 - 4 226 98
BC110511.1 - 3 - 4 226 98

View Full Product Details