The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024541.2 | NP_078817.2 | 23 - 24 | 1885 | 73 |
XM_006717969.1 | XP_006718032.1 | 21 - 22 | 1673 | 73 | |
XM_011540151.2 | XP_011538453.1 | 23 - 24 | 1930 | 73 | |
XM_011540152.2 | XP_011538454.1 | 22 - 23 | 1817 | 73 | |
XM_011540153.2 | XP_011538455.1 | 22 - 23 | 1781 | 73 | |
XM_011540154.2 | XP_011538456.1 | 21 - 22 | 1715 | 73 | |
XM_011540156.2 | XP_011538458.1 | 16 - 17 | 1849 | 73 | |
XM_011540157.2 | XP_011538459.1 | 22 - 23 | 1834 | 73 | |
XM_017016635.1 | XP_016872124.1 | 23 - 24 | 1883 | 73 | |
XM_017016636.1 | XP_016872125.1 | 23 - 24 | 1883 | 73 | |
GenBank mRNA | AK023176.1 | - | 6 - 7 | 514 | 73 |