The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105518.1 | NP_001098988.1 | 12 - 13 | 1954 | 112 |
NM_017673.6 | NP_060143.4 | 12 - 13 | 2005 | 112 | |
XM_005245271.1 | XP_005245328.1 | 12 - 13 | 2012 | 112 | |
XM_005245273.1 | XP_005245330.1 | 12 - 13 | 1913 | 112 | |
XM_011509657.1 | XP_011507959.1 | 12 - 13 | 1992 | 112 | |
XM_017001527.1 | XP_016857016.1 | 12 - 13 | 1892 | 112 | |
XM_017001528.1 | XP_016857017.1 | 12 - 13 | 2012 | 112 | |
XM_017001529.1 | XP_016857018.1 | 13 - 14 | 2217 | 112 | |
XM_017001530.1 | XP_016857019.1 | 12 - 13 | 2012 | 112 | |
GenBank mRNA | AF288392.1 | - | 11 - 12 | 1851 | 112 |
AK093263.1 | - | 6 - 7 | 717 | 112 | |
AK127980.1 | - | 12 - 13 | 1892 | 112 | |
AK291443.1 | - | 12 - 13 | 1871 | 112 | |
AK315802.1 | - | 12 - 13 | 1972 | 112 | |
BC030781.2 | - | 12 - 13 | 2007 | 112 |